Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD BBL™ Mycoflask™ Prepared Tubed Media: BHI Agar Slants with 5% Sheep Blood
Click to view available options
Quantity:
100/Pk.
For Use With (Application):
Isolation and cultivation of pathogenic and nonpathogenic fungi
Description
- For cultures requiring prolonged incubation

Specifications
Specifications
Certifications/Compliance | ISO/DIS, 11014, BS12, CMPH2, MCM9 |
Color | Dark Red |
Content And Storage | 2°C to 8°C |
Form | Liquid |
Manufacturing Origin | United States |
Media Type | BHI Agar |
Formulation | Brain heart infusion from (solids) 8g/L, Peptic digest of Animal Tissue 5g/L, Pancreatic digest of Casein 16g/L, Dextrose 2g/L, Sodium Chloride 5g/L, Disodium Phosphate 2.5g/L, Agar 13.5g/L |
For Use With (Application) | Isolation and cultivation of pathogenic and nonpathogenic fungi |
Packaging Type | Tube |
Quantity | 100/Pk. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction