Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Mycoflask™ Prepared Tubed Media: BHI Agar Slants with 5% Sheep Blood

Catalog No. p-4770198
Click to view available options
Quantity:
100/Pk.
For Use With (Application):
Isolation and cultivation of pathogenic and nonpathogenic fungi
This item is not returnable. View return policy
This item is not returnable. View return policy

Used in qualitative procedures for the isolation and cultivation of pathogenic and nonpathogenic fungi from clinical and nonclinical specimens

  • For cultures requiring prolonged incubation

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014, BS12, CMPH2, MCM9
Color Dark Red
Content And Storage 2°C to 8°C
Form Liquid
Manufacturing Origin United States
Media Type BHI Agar
Formulation Brain heart infusion from (solids) 8g/L, Peptic digest of Animal Tissue 5g/L, Pancreatic digest of Casein 16g/L, Dextrose 2g/L, Sodium Chloride 5g/L, Disodium Phosphate 2.5g/L, Agar 13.5g/L
For Use With (Application) Isolation and cultivation of pathogenic and nonpathogenic fungi
Packaging Type Tube
Quantity 100/Pk.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.