Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD BBL™ Mycoflask™ Sabouraud Dextrose Agar (Emmons) Slants, pH 6.9
Shop All BD Diagnostic Systems Products
Click to view available options
:
Sabouraud Dextrose Agar; 100/pk.
Description
Tubes with tight caps should be opened carefully to avoid injury due to breakage of glass.
Specifications
Specifications
| Certifications/Compliance | ISO/DIS, 11014, CMPH2, MCM9, SMWW |
| Color | Beige |
| Content And Storage | 2°C to 8°C |
| Form | Liquid |
| Manufacturing Origin | United States |
| Media Type | Sabouraud Dextrose Agar |
| Formulation | Pancreatic digest of Casein 5g/L, Peptic digest of Animal Tissue 5g/L, Dextrose 20g/L, Agar 17g/L |
| For Use With (Application) | Cultivation of dermatophyte, pathogenic and nonpathogenic fungi |
| Packaging Type | Tube with Tight Cap |
| Quantity | 100 Pack |
For in vitro diagnostic use.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction