Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Mycoflask™ Standard Methods Agar (APHA) Prepared Medai Deeps (Pour Tube)

Catalog No. bb21030 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
:
Standard methods agar (APHA) deeps; Pour tube; 20mL; Quantity: 10/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
BB21030 Standard methods agar (APHA) deeps; Pour tube; 20mL; Quantity: 10/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BB21030 Supplier BD Supplier No. 221030
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Used for obtaining microbial plate counts from milk and dairy products, foods, water and other materials of sanitary importance

Tubes with tight caps should be opened carefully to avoid injury due to breakage of glass.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014, AOAC, BAM, CCAM, COMPF, EPA, ISO, SMD, SMWW, USDA
Color Beige
Content And Storage 2 to 25°C
Form Liquid
Manufacturing Origin United States
Media Type Plate Count Agar
Formulation Pancreatic digest of Casein 5g/L, Yeast extract 2.5g/L, Dextrose 1g/L, Agar 15g/L
For Use With (Application) Used for obtaining microbial plate counts from Milk and dairy product, foods, Water and other materials of sanitary importance
Packaging Type Tube with Tight Cap
Quantity 20 mL

For in vitro diagnostic use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.