Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD BBL™ Mycoflask™ Thioglycollate (Fluid) Prepared Media
Click to view available options
Quantity:
10 x 8 mL
100 x 8 mL
100 x 20 mL
Color:
Beige
Light Beige
Media Type:
Thioglycollate (Fluid)
Description
This medium is capable of supporting good growth of a great variety of fastidious organisms, of both pathogenic and nonpathogenic species.
- Neutralize antibacterial activity of mercurial compounds
- Before use ∽30% of media should oxidized
Tubes with tight caps should be opened carefully to avoid injury due to breakage of glass.

Specifications
Specifications
Certifications/Compliance | CE, AOAC, BAM, BS12, COMPF, EP, USDA, USP |
Color | Light Beige |
Content And Storage | 2°C to 8°C |
Form | Liquid |
Manufacturing Origin | United States |
Media Type | Thioglycollate (Fluid) |
Formulation | Pancreatic digest of Casein 15g/L, L-Cystine 0.5g/L, Dextrose (anhydrous) 5g/L, Yeast extract 5g/L, Sodium Chloride 2.5g/L, Sodium Thioglycollate 0.5g/L, Resazurin 0.001g/L, Agar 0.75g/L |
For Use With (Application) | Sterility testing of biologics and for the cultivation of anaerobes, aerobes and microaerophiles |
Packaging Type | Tube |
Quantity | 10 x 8 mL |
For in vitro diagnostic use.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction