Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Mycoflask™ Thioglycollate (Fluid) Prepared Media

Catalog No. bb21196
Click to view available options
Quantity:
10 x 8 mL
100 x 8 mL
100 x 20 mL
Color:
Beige
Light Beige
Media Type:
Thioglycollate (Fluid)
This item is not returnable. View return policy
This item is not returnable. View return policy

Used for the sterility testing of biologics and for the cultivation of anaerobes, aerobes and microaerophiles

This medium is capable of supporting good growth of a great variety of fastidious organisms, of both pathogenic and nonpathogenic species.

  • Neutralize antibacterial activity of mercurial compounds
  • Before use ∽30% of media should oxidized

Tubes with tight caps should be opened carefully to avoid injury due to breakage of glass.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE, AOAC, BAM, BS12, COMPF, EP, USDA, USP
Color Light Beige
Content And Storage 2°C to 8°C
Form Liquid
Manufacturing Origin United States
Media Type Thioglycollate (Fluid)
Formulation Pancreatic digest of Casein 15g/L, L-Cystine 0.5g/L, Dextrose (anhydrous) 5g/L, Yeast extract 5g/L, Sodium Chloride 2.5g/L, Sodium Thioglycollate 0.5g/L, Resazurin 0.001g/L, Agar 0.75g/L
For Use With (Application) Sterility testing of biologics and for the cultivation of anaerobes, aerobes and microaerophiles
Packaging Type Tube
Quantity 100 x 8 mL

For in vitro diagnostic use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.