Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Mycoflask™ Trypticase™ Soy Agar (TSA II™) Deeps (Pour Tube) Prepared Media

Catalog No. b97941
Click to view available options
:
TS Agar (II) Deeps; 20mL Pour tubes; 100/pk

Used for cultivating fastidious microorganisms and for the visualization of hemolytic reactions produced by many bacterial species

Tubes with tight caps should be opened carefully to avoid injury due to breakage of glass.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE, BAM, COMPF
Color Beige
Content And Storage 2 to 25°C
Form Liquid
Manufacturing Origin United States
Media Type Trypticase Soy Agar (TSA II)
Formulation Pancreatic digest of Casein 14.5g/L, Papaic digest of Soybean Meal 5g/L, Sodium Chloride 5g/L, Agar 14g/L, Growth Factors 1.5g/L
For Use With (Application) Cultivation of fastidious microorganisms and visualization of hemolytic reactions
Packaging Type Tube with Tight Cap
Quantity 20 mL

For in vitro diagnostic use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.