Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Prepared Media Chocolate II Agar Slants

Catalog No. l95872 Shop All BD Diagnostic Systems Products
Click to view available options
:
Chocolate II Agar Slants; K size tubes; 10/Pk.
This item is not returnable. View return policy
This item is not returnable. View return policy

Requires

Ancillary culture media, reagents, quality control organisms and laboratory equipment

Enriched medium for cultivation of Neisseria and Haemophilus species

Tubes with tight caps should be opened carefully to avoid injury due to breakage of glass.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE, BS12, CMPH2, MCM9
Color Dark Brown
Content And Storage 2°C to 8°C
Form Liquid
Manufacturing Origin United States
Media Type Chocolate Agar
Formulation Pancreatic digest of Casein 7.5g/L, Selected meat Peptone 7.5g/L, Corn Starch 1g/L, Dipotassium Phosphate 4g/L, Monopotassium Phosphate 1g/L, Sodium Chloride 5g/L, Agar 12g/L, Hemoglobin 10g/L, IsoVitaleX™ enrichment 10mL
For Use With (Application) Cultivation of Neisseria and Haemophilus species
Packaging Type Tube
Quantity 10/Pk.

For in vitro diagnostic use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.