Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Prepared Plated Media: Bacteroides Bile Esculin Agar (BBE)

Catalog No. l21836
Click to view available options
:
Bacteroides Bile Esculin Agar (BBE); 10/Pk.
This item is not returnable. View return policy
This item is not returnable. View return policy

For primary isolation medium for the selection and presumptive identification of the B. fragilis group

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014
Color Light Beige
Form Liquid
Manufacturing Origin United States
Media Type Bacteroides Bile Esculin Agar
Formulation Pancreatic digest of Casein 14.5g/L, Papaic digest of Soybean Meal 5g/L, Sodium Chloride 5g/L, Esculin 1g/L, Ferric Ammonium Citrate 0.5g/L, Oxgall 15g/L, Hemin 0.01g/L, Gentamicin 0.1g/L, Vitamin K1 0.01g/L, Agar 14g. Growth Factors 1.8g/L
For Use With (Application) Primary isolation medium for the selection and presumptive identification of the B. fragilis group
Packaging Type Plate
Quantity 10/Pk.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.