Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Prepared Plated Media: Baird-Parker Agar

Catalog No. B4397214X
Click to view available options
Quantity:
100/Pk.
20/Pk.
Color:
Light Beige
Red
This item is not returnable. View return policy
This item is not returnable. View return policy

For isolation and presumptive identification of coagulase-positive staphylococci (Staphylococcus aureus) from clinical specimens, foods, air, water and other materials

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014
Color Red
Content And Storage 2°C to 8°C
Form Liquid
Manufacturing Origin United States
Media Type Baird Parker Agar
Formulation Pancreatic digest of Casein 10g/L, Beef extract 5g/L, Yeast extract 1g/L, Lithium Chloride 5g/L, Agar 17g/L, Glycine 12g/L, Sodium Pyruvate 10g/L, Potassium Tellurite solution, 1% 1mL, Egg Yolk Emulsion, 50% 50mL
For Use With (Application) Coagulase positive Staphylococci
Packaging Type Plate
Quantity 20/Pk.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.