Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD BBL™ Prepared Plated Media: Baird-Parker Agar
Shop All BD Diagnostic Systems Products
Click to view available options
Quantity:
100/Pk.
20/Pk.
Color:
Light Beige
Red
Specifications
Specifications
Certifications/Compliance | ISO/DIS, 11014 |
Color | Red |
Content And Storage | 2°C to 8°C |
Form | Liquid |
Manufacturing Origin | United States |
Media Type | Baird Parker Agar |
Formulation | Pancreatic digest of Casein 10g/L, Beef extract 5g/L, Yeast extract 1g/L, Lithium Chloride 5g/L, Agar 17g/L, Glycine 12g/L, Sodium Pyruvate 10g/L, Potassium Tellurite solution, 1% 1mL, Egg Yolk Emulsion, 50% 50mL |
For Use With (Application) | Coagulase positive Staphylococci |
Packaging Type | Plate |
Quantity | 20/Pk. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction