Learn More
BD BBL™ Prepared Plated Media: CDC Anaerobe 5% Sheep Blood Agar with Phenylethyl Alcohol (PEA)
Shop All BD Diagnostic Systems ProductsDescription
- Ready-to-use
- 100 x 10mm and 100 x 15mm-style plates
Specifications
Specifications
| Certifications/Compliance | ISO, 9000 |
| Color | Red |
| Form | Liquid |
| Manufacturing Origin | United States |
| Media Type | CDC anaerobe Blood Agar with Phenylethyl Alcohol |
| Formulation | Pancreatic digest of Casein 15g/L, Papaic digest of Soybean Meal 5g/L, Sodium Chloride 5g/L, Agar 20g/L, Yeast extract 5g/L, Hemin 0.005g/L, Vitamin K1 0.01g/L, L-Cystine 0.4g/L, Phenylethyl Alcohol 2.5g/L, Sheep Blood, defibrinated 5% |
| For Use With (Application) | Isolation of fastidious and slow growing, obligately anaerobic bacteria from a variety of clinical and nonclinical materials |
| Packaging Type | Plate |
| Quantity | 20/Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.