Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Prepared Plated Media: CDC Anaerobe Laked Sheep Blood Agar w/Kanamycin and Vancomycin (KV)

Catalog No. b21846x Shop All BD Diagnostic Systems Products
Click to view available options
:
CDC Anaerobe Laked Sheep Blood Agar wiith Kanamycin and Vancomycin (KV); 20/Pk.
This item is not returnable. View return policy
This item is not returnable. View return policy

For the selective isolation of fastidious and slow-growing, obligately anaerobic bacteria from a variety of clinical and nonclinical materials

  • Ready-to-use
  • 100 x 10mm and 100 x 15mm-style plates

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014
Color Dark Red
Content And Storage 2°C to 8°C
Form Liquid
Manufacturing Origin United States
Media Type CDC anaerobe Laked Sheep Blood Agar with Kanamycin and Vancomycin
Formulation Pancreatic digest of Casein 15g/L, Papaic digest of Soybean Meal 5g/L, Sodium Chloride 5g/L, Agar 20g/L, Yeast extract 5g/L, Hemin 0.005g/L, Vitamin K1 0.01g/L, L-Cystine 0.4g/L, Kanamycin 0.1g/L, Vancomycin 0.0075g/L, Sheep Blood, defibrinated, laked 5%
For Use With (Application) Fastidious and slow growing obligately anaerobic gram negative bacteria
Packaging Type Plate
Quantity 20/Pk.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.