Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Prepared Plated Media: Chocolate II Agar (GC II Agar with Hemoglobin and IsoVitaleX™)

Catalog No. p-4766067
Click to view available options
Quantity:
100/Pk.
20/Pk.
This item is not returnable. View return policy
This item is not returnable. View return policy

For the isolation and cultivation of fastidious microorganisms, especially Neisseria and Haemophilus species, from a variety of clinical specimens

  • Ready-to-use
  • 100 x 10mm and 100 x 15mm-style plates

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014
Color Dark Brown
Content And Storage 2°C to 8°C
Form Agar
Manufacturing Origin United States
Media Type Chocolate II Agar (GC II Agar with Hemoglobin and IsoVitaleX™)
Formulation Pancreatic digest of Casein 7.5g/L, Selected meat Peptone 7.5g/L, Corn Starch 1g/L, Dipotassium Phosphate 4g/L, Monopotassium Phosphate 1g/L, Sodium Chloride 5g/L, Agar 12g/L, Hemoglobin 10g/L, IsoVitaleX™ enrichment 10mL
For Use With (Application) Neisseria and Haemophilus species
Packaging Type Plate
Quantity 100/Pk.
Weight (Metric) 3800 g
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.