Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD BBL™ Prepared Plated Media: CHROMagar™ Salmonella
Click to view available options
:
CHROMagar Salmonella; 20/Pk.
Description
- Ready-to-use
- 100 x 10mm and 100 x 15mm-style plates

Specifications
Specifications
Certifications/Compliance | ISO/DIS, 11014 |
Color | Beige |
Content And Storage | 2°C to 8°C |
Form | Liquid |
Manufacturing Origin | United States |
Media Type | Chromagar Salmonella |
Formulation | Chromopeptone 22g/L, Chromogenic Mix 0.34g/L, Inhibitory Agents 0.02g/L, Agar 15g/L |
For Use With (Application) | Salmonella species from other Coliform and non-coliform bacteria in clinical stool samples and a variety of food samples |
Packaging Type | Plate |
Quantity | 20/Pk. |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction