Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD BBL™ Prepared Plated Media: GC-Lect™ Agar
Shop All BD Diagnostic Systems Products
Click to view available options
Quantity:
100/Pk.
Weight (Metric):
4000 g
Description
Selective media provide inhibition of contaminating bacteria and fungi, including Capnocytophaga species in oropharyngeal specimens.
- Ready-to-use
- 100 x 10mm and 100 x 15mm-style plates

Specifications
Specifications
Certifications/Compliance | ISO/DIS, 11014 |
Color | Dark Brown |
Form | Liquid |
Manufacturing Origin | United States |
Media Type | GC-Lect™ Agar |
Formulation | Pancreatic digest of Casein 7.5g/L, Selected meat Peptone 7.5g/L, Corn Starch 1g/L, Dipotassium Phosphate 4g/L, Monopotassium Phosphate 1g/L, Sodium Chloride 5g/L, Agar 12g/L, Hemoglobin 10g/L, selective Agents 17mg/L, IsoVitaleX™ enrich |
For Use With (Application) | Neisseria gonorrhoeae |
Packaging Type | Plate |
Quantity | 100/Pk. |
Weight (Metric) | 4000 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction