Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Prepared Plated Media: Hektoen Enteric Agar

Catalog No. p-4766069
Click to view available options
Quantity:
100/Pk.
20/Pk.
Weight (Metric):
1170 g
3000 g
This item is not returnable. View return policy
This item is not returnable. View return policy

For the isolation and cultivation of gram-negative enteric microorganisms, especially Shigella, from a variety of clinical and nonclinical specimens

  • Ready-to-use
  • 100 x 10mm and 100 x 15mm-style plates

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014
Color Light Beige
Form Solid
Manufacturing Origin United States
Media Type Hektoen enteric Agar
Formulation Peptic digest of Animal Tissue 12g/L, Yeast extract 3g/L, Bile Salts 9g/L, Lactose 12g/L, Sucrose 12g/L, Salicin 2g/L, Sodium Chloride 5g/L, Sodium Thiosulfate 5g/L, Ferric Ammonium Citrate 1.5g/L, Bromthymol blue 0.064g/L, Acid Fuchsin 0.1g/L, Agar 13.5g/L
For Use With (Application) Gram negative enteric microorganisms, especially Shigella
Packaging Type Plate
Quantity 100/Pk.
Weight (Metric) 3000 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.