Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Prepared Plated Media: MacConkey II Agar with Sorbitol

Catalog No. p-4766458 Shop All BD Diagnostic Systems Products
Click to view available options
Quantity:
10/Pk.
100/Pk.
Media Type:
MacConkey II Agar
MacConkey II Agar with Sorbitol
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective and differential media for the detection of Escherichia coli serotype O157:H7 associated with hemorrhagic colitis

The media is also used for the isolation of O157:H7 from foods.

  • Ready-to-use
  • Contains chloramphenicol
  • 100 x 10mm and 100 x 15mm-style plates
  • Deep-filled plates reduce effects of drying during prolonged incubation

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014
Color Red
Content And Storage 2°C to 8°C
Form Liquid
Manufacturing Origin United States
Media Type MacConkey II Agar with Sorbitol
Formulation Pancreatic digest of Gelatin 17g/L, Pancreatic digest of Casein 1.5g/L, Peptic digest of Animal Tissue 1.5g/L, D-Sorbitol 10g/L, Bile Salts 1.5g/L, Sodium Chloride 5g/L, Neutral red 0.03g/L, Crystal Violet 0.001g/L, Agar 13.5g/L
For Use With (Application) Detection of Escherichia coli serotype O157:H7 associated with hemorrhagic colitis
Packaging Type Plate
Quantity 100/Pk.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.