Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD BBL™ Prepared Plated Media: Mannitol Salt Agar
Shop All BD Diagnostic Systems Products
Click to view available options
Quantity:
100/Pk.
20/Pk.
Certifications/Compliance:
ISO, 9000
USP, ISO/DIS, 11014
For Use With (Application):
Staphylococci detection/count
Description
- Ready-to-use
- 100 x 10mm and 100 x 15mm-style plates
Specifications
Specifications
| Certifications/Compliance | USP, ISO/DIS, 11014 |
| Color | Red |
| Content And Storage | 2°C to 8°C |
| Form | Liquid |
| Manufacturing Origin | United States |
| Media Type | Mannitol Salt Agar |
| Formulation | Beef extract 1g/L, Pancreatic digest of Casein 5g/L, Peptic digest of Animal Tissue 5g/L, Sodium Chloride 75g/L, D-Mannitol 10g/L, Phenol red 0.025g/L, Agar 15g/L |
| For Use With (Application) | Staphylococci detection/count |
| Packaging Type | Plate |
| Quantity | 20/Pk. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction