Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD BBL™ Prepared Plated Media: Middlebrook and Cohn 7H10 Agar (Deep Fill)
Click to view available options
:
Middlebrook and Cohn 7H10 agar (deep fill); 20/Pk.
Description
- Ready-to-use
- 100 X 10mm and 100 X 15mm style plates
- Deep-filled plates reduce effects of drying during prolonged incubation

Specifications
Specifications
Certifications/Compliance | ISO/DIS, 11014 |
Color | Light Green |
Form | Liquid |
Manufacturing Origin | United States |
Media Type | Middlebrook and Cohn 7H10 agar (deep fill) |
Formulation | Magnesium Sulfate 0.05g/L, Ferric Ammonium Citrate 0.04g/L, Sodium Citrate 0.4g/L, Ammonium Sulfate 0.5g/L, Monosodium Glutamate 0.5g/L, Disodium Phosphate 1.5g/L, Monopotassium Phosphate 1.5g/L, Agar 13.5g/L, Sodium Chloride 0.85g/L, Dextrose 2g/L, Bovine Albumin V 5g/L, Catalase 3mg/L, Pyridoxine 1mg/L, Zinc Sulfate 1mg/L, Copper Sulfate 1mg/L, Biotin 0.5mg/L, Calcium Chloride 0.5mg/L, Malachite green 0.25mg/L, Oleic Acid 0.06mL, Glycerol 5mL |
For Use With (Application) | Mycobacteria |
Packaging Type | Plate |
Quantity | 20/Pk. |
Weight (Metric) | 1380 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction