Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Prepared Plated Media: Mueller Hinton Agar with 5% Sheep Blood

Catalog No. p-4766428
Click to view available options
Quantity:
20/Pk.
24/Pk.
This item is not returnable. View return policy
This item is not returnable. View return policy

For antimicrobial disc diffusion susceptibility testing of Streptococcus pneumoniae with selected agents

  • Ready-to-use
  • 100 X 10mm, 100 X 15mm and 150 x 15mm-style plates

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014
Color Red
Form Solid
Manufacturing Origin United States
Media Type Mueller Hinton Agar with 5% Sheep Blood
Formulation Beef extract 2g/L, Acid Hydrolysate of Casein 17.5g/L, Starch 1.5g/L, Agar 17g/L, Sheep Blood, defibrinated 5%
For Use With (Application) Antimicrobial disc diffusion susceptibility testing of Streptococcus pneumoniae with selected agents
Packaging Type Plate
Quantity 20/Pk.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.