Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD BBL™ Prepared Plated Media: Mycosel™ Agar (Deep Fill)
Click to view available options
:
Mycosel agar (deep fill); 20/Pk.
Description
- Ready-to-use
- Contains cycloheximide and chloramphenicol
- 100 X 10mm and 100 X 15mm style plates
- Deep-filled plates reduce effects of drying during prolonged incubation

Specifications
Specifications
Certifications/Compliance | ISO/DIS, 11014 |
Color | Light Yellow |
Content And Storage | 2°C to 8°C |
Form | Liquid |
Manufacturing Origin | United States |
Media Type | Mycosel agar |
Formulation | Papaic digest of Soybean Meal 10g/L, Dextrose 10g/L, Agar 15.5g/L, Cycloheximide 0.4g/L, Chloramphenicol 0.05g/L |
For Use With (Application) | Pathogenic fungi |
Packaging Type | Plate |
Quantity | 20/Pk. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction