Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD BBL™ Prepared Plated Media: Selective Streptococcus Agar
Shop All BD Diagnostic Systems Products
Click to view available options
Quantity:
20/Pk.
Description
- Ready-to-use
- 100 x 10mm and 100 x 15mm-style plates

Specifications
Specifications
Certifications/Compliance | ISO/DIS, 11014 |
Color | Light Beige |
Content And Storage | 2°C to 8°C |
Form | Liquid |
Manufacturing Origin | United States |
Media Type | Selective Streptococcus Agar |
Formulation | Pancreatic digest of Casein 10g/L, Beef extract 6.7g/L, Sodium Chloride 5g/L, Maltose 0.25g/L, Agar 15g/L, Nucleic Acid 6g/L, Neomycin Sulfate 0.002g/L, Polymyxin B Sulfate 200,000U, Sheep Blood, defibrinated 5% |
For Use With (Application) | Group A streptococci from respiratory sources |
Packaging Type | Plate |
Quantity | 20/Pk. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction