Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Prepared Plated Media: Trypticase™ Soy Agar (Soybean-Casein Digest Agar)

Catalog No. b21283x Shop All BD Diagnostic Systems Products
Click to view available options
Quantity:
100/Pk.
Media Type:
Trypticase Soy Agar
This item is not returnable. View return policy

Catalog No. B21283X

Supplier: BD 221283

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Used for isolation and cultivation of non-fastidious and fastidious microorganisms

  • Ready for immediate use
  • Not used for anaerobes

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014
Color Light Beige
Content And Storage 2°C to 8°C
Manufacturing Origin United States
Media Type Trypticase Soy Agar
Formulation Pancreatic digest of Casein 15g/L, Papaic digest of Soybean Meal 5g/L, Sodium Chloride 5g/L, Agar 15g/L
For Use With (Application) Cultivation of Non-fastidious and fastidious microorganisms (not for anaerobes)
Packaging Type Plate
Quantity 100/Pk.
Dimensions (Dia. x H) 100 x 15 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.