Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD BBL™ Prepared Plated Media: Trypticase™ Soy Agar (Soybean-Casein Digest Agar)
Shop All BD Diagnostic Systems Products
Click to view available options
Quantity:
100/Pk.
20/Pk.
Media Type:
Trypticase Soy Agar
Description
- Ready for immediate use
- Not used for anaerobes

Specifications
Specifications
Certifications/Compliance | ISO/DIS, 11014 |
Color | Light Beige |
Content And Storage | 2°C to 8°C |
Manufacturing Origin | United States |
Media Type | Trypticase Soy Agar |
Formulation | Pancreatic digest of Casein 15g/L, Papaic digest of Soybean Meal 5g/L, Sodium Chloride 5g/L, Agar 15g/L |
For Use With (Application) | Cultivation of Non-fastidious and fastidious microorganisms (not for anaerobes) |
Packaging Type | Plate |
Quantity | 100/Pk. |
Dimensions (Dia. x H) | 100 x 15 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction