Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Prepared Plated Media: V Agar

Catalog No. p-4766868 Shop All BD Diagnostic Systems Products
Click to view available options
Quantity:
10/Pk.
100/Pk.
This item is not returnable. View return policy
This item is not returnable. View return policy

Nonselective enriched media used in qualitative procedures for the isolation and differentiation of Gardnerella vaginalis from clinical specimens

  • Ready-to-use
  • Enriched with Columbia agar base supplemented with 5% human blood
  • 100 x 10mm and 100 x 15mm-style plates

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014
Color Light Beige
Content And Storage 2°C to 8°C
Form Liquid
Manufacturing Origin United States
Media Type V Agar (for G. vaginalis)
Formulation Pancreatic digest of Casein 12g/L, Peptic digest of Animal Tissue 5g/L, Beef extract 3g/L, Yeast extract 3g/L, Peptone 10g/L, Corn Starch 1g/L, Sodium Chloride 5g/L, Agar 13.5g/L, Human Blood, anticoagulated 5%
For Use With (Application) Gardnerella vaginalis from clinical specimens
Packaging Type Plate
Quantity 10/Pk.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.