Learn More
Description
- Sterile
- Contains lecithin and polysorbate 80
Specifications
Specifications
| Certifications/Compliance | ISO/DIS, 11014 |
| Color | Light Beige |
| Content And Storage | 2°C to 8°C |
| Form | Liquid |
| Manufacturing Origin | United States |
| Media Type | Sabouraud Dextrose Agar with Lecithin and Polysorbate 80 |
| Sterility | Sterile |
| Packaging Type | Sterile Rodac™ Plate |
| Quantity | 100/Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.