Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD BBL™ Sabouraud Dextrose Agar Mycoflask™ Prepared Media
Click to view available options
:
Sabouraud dextrose agar Mycoflask; Quantity 10
Description
Tubes with tight caps should be opened carefully to avoid injury due to breakage of glass.

Specifications
Specifications
Certifications/Compliance | ISO/DIS, 11014, BAM, CCAM, CMPH2, COMPF, EP, JP, MCM9, USP |
Color | Yellow Brown |
Content And Storage | 2°C to 8°C |
Form | Liquid |
Manufacturing Origin | United States |
Media Type | Sabouraud Dextrose Agar |
Formulation | Pancreatic digest of Casein 5g/L, Peptic digest of Animal Tissue 5g/L, Dextrose 40g/L, Agar 15g/L |
For Use With (Application) | Cultivation of dermatophytes |
Packaging Type | Mycoflask™ Bottle |
Quantity | 10/Pk. |
For in vitro diagnostic use
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction