Learn More
BD BBL™ Sensi-Disc™ Antimicrobial Susceptibility Test Discs, Aztreonam8
Shop All BD Diagnostic Systems Products
                    Click to view available options
                                Quantity:
                                10 Pack
                                Each
                    Description
- 6mm discs impregnated with accurately determined amounts of antibiotic or chemotherapeutic agents
- Are clearly marked on both sides with letters and numbers designating agent and drug content
            Specifications
Specifications
| Antimicrobial Agents | Aztreonam8 | 
| Concentration | 30 μg | 
| Includes | 50 discs/cartridge | 
| Packaging Type | Disc | 
| Quantity | Each | 
For in vitro diagnostic use