Learn More
BD BBL™ Sensi-Disc™ Antimicrobial Susceptibility Test Discs, Cefotaxime/Clavulanic Acid
Catalog No. p-4638877 Shop All BD Diagnostic Systems Products
Click to view available options
Quantity:
1 Pack
10 Pack
This item is not returnable. View return policy
Catalog No. B231751
Supplier: BD 231751
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Used for semi-quantitative in vitro testing by agar disc diffusion test procedure of certain fastidious bacterial pathogens
  • 6mm discs impregnated with accurately determined amounts of antibiotic or chemotherapeutic agents
  • Are clearly marked on both sides with letters and numbers designating agent and drug content
TRUSTED_SUSTAINABILITY
Specifications
Antimicrobial Agents Cefotaxime/Clavulanic Acid
Concentration 10 μg
Packaging Type Disc
Quantity 1 Pack
For in vitro diagnostic use
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.