Learn More
BD BBL™ Sensi-Disc™ Antimicrobial Susceptibility Test Discs, Linezolid85
Shop All BD Diagnostic Systems ProductsDescription
- 6mm discs impregnated with accurately determined amounts of antibiotic or chemotherapeutic agents
- Are clearly marked on both sides with letters and numbers designating agent and drug content
Specifications
Specifications
| Antimicrobial Agents | Linezolid 85 |
| Concentration | 30 μg |
| Packaging Type | Disc |
| Quantity | 10 Pack |
For in vitro diagnostic use