Learn More
BD BBL™ Sensi-Disc™ Susceptibility Test Discs: Doripenem DOR-10
Catalog No. b232219 Shop All BD Diagnostic Systems Products
Click to view available options
Quantity:
10 Pack
This item is not returnable. View return policy
This item is not returnable. View return policy
High-quality absorbent paper discs impregnated with accurately determined amounts of antibiotic or other chemotherapeutic agents
  • Discs containing certifiable agents are submitted to FDA for assay and certification before distribution Clearly marked on both sides with letters and numbers designating agent and concentration
TRUSTED_SUSTAINABILITY
Specifications
Antimicrobial Agents Doripenem DOR-10
Packaging Type Disc
Quantity 10 Pack
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.