Learn More
BD BBL™ Sensi-Disc™ Susceptibility Test Discs: Furazolidone
Shop All BD Diagnostic Systems Products
Click to view available options
Concentration:
100 μg
Description
- Discs containing certifiable agents are submitted to FDA for assay and certification before distribution Clearly marked on both sides with letters and numbers designating agent and concentration
Specifications
Specifications
Antimicrobial Agents | Furazolidone |
Concentration | 100 μg |
Packaging Type | Disc |
Quantity | 50 Discs |