Learn More
BD BBL™ Sensi-Disc™ Susceptibility Test Discs: Nalidixic Acid
Catalog No. p-4899838 Shop All BD Diagnostic Systems Products
Click to view available options
Quantity:
1/Pk.
10/Pk.
This item is not returnable. View return policy
This item is not returnable. View return policy
High-quality absorbent paper discs impregnated with accurately determined amounts of antibiotic or other chemotherapeutic agents
  • Discs containing certifiable agents are submitted to FDA for assay and certification before distribution Clearly marked on both sides with letters and numbers designating agent and concentration
TRUSTED_SUSTAINABILITY
Specifications
Antimicrobial Agents Nalidixic Acid
Concentration 30 μg
Packaging Type Disc
Quantity 1/Pk.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.