Learn More
BD BBL™ Sensi-Disc™ Susceptibility Test Discs: Neomycin (5μg)
Shop All BD Diagnostic Systems Products
Click to view available options
Concentration:
5 μg
Description
- Discs containing certifiable agents are submitted to FDA for assay and certification before distribution Clearly marked on both sides with letters and numbers designating agent and concentration
Specifications
Specifications
| Antimicrobial Agents | Neomycin |
| Concentration | 5 μg |
| Packaging Type | Disc |
| Quantity | 10 Pack |