Learn More
BD BBL™ Sensi-Disc™ Susceptibility Test Discs: Piperacillin
Catalog No. p-4897824 Shop All BD Diagnostic Systems Products
Click to view available options
Quantity:
1/Pk.
10/Pk.
This item is not returnable. View return policy
This item is not returnable. View return policy
High-quality absorbent paper discs impregnated with accurately determined amounts of antibiotic or other chemotherapeutic agents
  • Clearly marked on both sides with letters and numbers designating agent and concentration
  • Available in pack of 1 and 10
TRUSTED_SUSTAINABILITY
Specifications
Antimicrobial Agents Piperacillin
Concentration 100 μg
Packaging Type Disc
Quantity 10/Pk.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.