Learn More
Description
Includes
BBL Staphyloslide Test Latex; BBL Staphyloslide Control Latex; Test cards, Mixing sticks
The BD BBL Staphyloslide Latex Test Kit utilizes blue polystyrene latex particles that have been sensitized with fibrinogen and IgG. When Staphylococcal colonies which possess bound coagulase (clumping factor) and/or protein A are mixed with the latex reagent, the latex particles will agglutinate strongly within 20 seconds.
Recommended Storage
Store at 2° to 8°C. Do not freeze.
Specifications
Specifications
| Content And Storage | 2°C to 8°C Do not freeze |
| Description | BBL Staphyloslide Latex Test Kit |
| Detectable Analytes | S. aureus |
| For Use With (Application) | Staphylococcus aureus Testing |
| Includes | BBL Staphyloslide Test Latex, 2 x 2.5mL; BBL Staphyloslide Control Latex, 2.5mL; Test cards, Mixing sticks |
| Quantity | 100 Tests |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.