BD BBL™ Sterile Finger Dab Plate: Trypticase Soy Agar w/Lecithin and Polysorbate 80

Plated media in gamma-irradiated packs for isolator systems multiwrap packaging protects media from vaporized hydrogen peroxide exposure

Manufacturer: BD 292271

 View more versions of this product

Catalog No. 22-285-110

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Specifications

    Sterile Yes
    Format 150 x 15mm Finger Dab
    Quantity 100/Pk.
    Color Light Beige
    Manufacturing Origin United States
    Media Type Trypticase™ Soy Agar with Lecithin and Polysorbate 80
    Packaging Plate
    Shelf Life 154 Days
    Certifications/Compliance ISO/DIS, 11014

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit