Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD BBL™ Sterile Finger Dab Plate: Trypticase Soy Agar w/Lecithin and Polysorbate 80
missing translation for 'orderingAttributeHoverText'
Quantity:
100/Pk.
5/Pk.
Certifications/Compliance:
ISO/DIS, 11014
USP
Description
- Available in 100 x 15mm Style Plates

Specifications
Specifications
Certifications/Compliance | USP |
Color | Light Beige |
Content And Storage | 2°C to 8°C |
Format | 150 x 15mm Finger Dab |
Manufacturing Origin | United States |
Media Type | Trypticase™ Soy Agar with Lecithin and Polysorbate 80 |
Sterility | Sterile |
For Use With (Application) | Sterility testing and process validation with media that are able to support a wide range of microorganisms |
Includes | Inner bag for transportation |
Packaging Type | Plate |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction