Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD BBL™ Trypticase™ Soy Agar w/5% Sheep Blood/ Chocolate II, I Plate™ Prepared Media
Click to view available options
Quantity:
100/Pk.
Media Type:
Trypticase™ Soy Agar with 5% Sheep Blood (TSA II) and chocolate II Agar (GC II Agar with hemoglobin and IsoVitaleX™)
For Use With (Application):
Cultivation of fastidious microorganisms and for visualization of hemolytic reaction
Description
Tripticase Soy Agar (TSA II)
- For the visualization of hemolytic reactions produced by many bacterial species
- GC II Agar with Hemoglobin and IsoVitaleX™
- For use in qualitative procedures for isolation and cultivation of fastidious microorganisms, Neisseria and Haemophilus species

Specifications
Specifications
Certifications/Compliance | ISO/DIS, 11014 |
Color | Dark Brown |
Form | Liquid |
Manufacturing Origin | United States |
Media Type | Trypticase™ Soy Agar with 5% Sheep Blood (TSA II) and chocolate II Agar (GC II Agar with hemoglobin and IsoVitaleX™) |
Formulation | Pancreatic digest of Casein 14.5g/L, Papaic digest of Soybean Meal 5g/L, Sodium Chloride 5g/L, Agar 14g/L, Growth Factors 1.5g/L, Defibrinated Sheep Blood 5%, Pancreatic digest of Casein 7.5g/L, Selected meat Peptone 7.5g/L, Corn Starch 1g/L, Dipotassium |
For Use With (Application) | Cultivation of fastidious microorganisms and for visualization of hemolytic reaction |
Packaging Type | Plate |
Quantity | 100/Pk. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction