Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Trypticase™ Soy Agar w/5% Sheep Blood/Levine EMB Agar, I Plate™ Prepared Media

Catalog No. p-4770155 Shop All BD Diagnostic Systems Products
Click to view available options
Quantity:
100/Pk.
This item is not returnable. View return policy
This item is not returnable. View return policy

Two media formulations in single plate for cultivating fastidious microorganisms

Tripticase Soy Agar (TSA II)

  • For the visualization of hemolytic reactions produced by many bacterial species
Levine EMB Agar
  • Selective and differential plating medium for isolation of gram-negative enteric bacteria

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014
Color Dark Yellow
Form Liquid
Manufacturing Origin United States
Media Type Trypticase™ Soy Agar with 5% Sheep Blood (TSA II)/Levine EMB Agar
For Use With (Application) Cultivation of fastidious microorganisms and for visualization of hemolytic reaction
Packaging Type Plate
Quantity 100/Pk.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.