Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Vacutainer™ Anaerobic Specimen Collector

Catalog No. l4336500 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
100/Cs.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L4336500 100/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. L4336500 Supplier BD Supplier No. 236500
This item is not returnable. View return policy

The only media-free device for anaerobic specimen collection available.

Media-free specimen collector helps to minimize sample dilution which may occur in media-based products and eliminates the potential for non-viables that can be seen in some transport media. Comfortable to use, it keeps anaerobic bacteria viable for up to 72 hours. The device offers a built-in oxygen-elimination system that is activated by depressing the plunger and includes a reliable color-change indicator to ensure that anaerobiasis has been achieved.

  • Sterile collector with swab provides a method to protect anaerobic bacteria from exposure to toxic amounts of oxygen during the time from specimen collection to inoculation of suitable media
  • The system may also be used for liquid specimens collected with a needle and syringe
  • Although primarily designed for the collection of specimens suspected of containing anaerobic organisms, aerobic organisms will survive in this system
  • Particularly suited for instances where volumes are small or dilution of specimens by a transport medium is to be avoided
  • Provides a moist, anaerobic atmosphere
  • Although prolonged storage of any specimen is not recommended, provides 72 hours (or longer) survival of anaerobes
  • Sterile collectors are packaged in peel-apart envelopes perfect for operating room use

TRUSTED_SUSTAINABILITY

Specifications

Product Type Anaerobic Specimen Collector
Sterility Sterile
For Use With (Application) For the collection of specimens suspected of containing anaerobic organisms
Quantity 100/Cs.
Storage Requirements 4°C to 25°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.