Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ VCNT Inhibitor (Thayer and Martin)

Catalog No. b12408
Click to view available options
:
VCNT Inhibitor (Thayer and Martin); BBL; 10mL
This item is not returnable. View return policy
This item is not returnable. View return policy

For selection of pathogenic Neisseria, Thayer-Martin Agar and Transgrow medium

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014
Additive VCNT Inhibitor
Form Lyophilized
For Use With (Application) Selection of pathogenic Neisseria, Thayer-Martin Agar and Transgrow medium
Includes 300μg Vancomycin, 750μg Colistin, 1200U Nystatin, 500μg Trimethoprim Lactate
Packaging Type Bottle
Quantity 10 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.