Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BD Biosciences Fixable Viability Stain 620

Catalog No. bdb564996 Shop All BD Biosciences Products
Click to view available options
Quantity:
100 μg

For discrimination of viable from non-viable mammalian cells in multicolor flow cytometric applications

  • Reacts with and covalently binds to cell-surface and intracellular amines
  • Permeable plasma cell membranes, such as those present in necrotic cells, allow intracellular diffusion of dye and covalent binding to higher overall concentrations of amines than in non-permeable live cells
  • Necrotic cells present in a typical in vitro assay label with higher levels of dye increases their fluorescence intensity 10-20 fold over viable cells
  • Labeled cells can be fixed with formaldehyde for downstream decontamination, freezing and/or permeabilization and subsequent intracellular staining while maintaining stable viability stain fluorescence
  • Excited by Yellow-Green laser (with an excitation maximum of 523nm), but is also well-excited by the Blue laser
  • Fluorescence emission maximum of 617nm
  • Applications: Flow cytometry, Intracellular staining
TRUSTED_SUSTAINABILITY

Specifications

Quantity 100 μg
For Use With (Application) Flow cytometry, Intracellular staining (flow cytometry) (Tested During Development)

RUO

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.