Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Biosciences Transcription Factor Buffer Set, BD
Shop All BD Biosciences Products
Click to view available options
Quantity:
Ea.
Description
Transcription Factor Buffer Set was designed to improve ease-of-use and minimize processing time, to reduce nonspecific staining, to increase the resolution of positively stained cells and to significantly reduce cell loss during fixation, permeabilization and staining procedures.
- Contents: Fixation/permeabilization buffer, diluent buffer, perm/wash buffer
Specifications
Specifications
| Chemical Name or Material | Transcription factor Buffer set |
| For Use With (Application) | Intracellular block, flow cytometry, staining of transcription factors alone or in combination with cell surface markers and cytokines |
| Quantity | Ea. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction