Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD Bottled Media Saboraud Dextrose Agar w/Lecithin, Polysorbate 80
Click to view available options
:
10/Pk.
Description
- With Lecithin and Polysorbate 80

Specifications
Specifications
Color | Light Beige |
Form | Liquid |
Manufacturing Origin | United States |
Formulation | Pancreatic digest of Casein 5g/L, Peptic digest of Animal Tissue 5g/L, Dextrose 40g/L, Agar 15g/L |
For Use With (Application) | Detection and enumeration of microorganisms present on surfaces of sanitary importance |
Packaging Type | Bottle |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction