Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Bottled Media TAT Broth

Catalog No. 14959800
Click to view available options
Quantity:
90 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

For sterility and non-sterile product testing

  • For cultivating microorganisms from highly-viscous or gelatinous materials
  • For testing of cosmetics

TRUSTED_SUSTAINABILITY

Specifications

Form Liquid
Manufacturing Origin United States
Fill Volume 90 mL
Formulation Pancreatic digest of Casein 20g/L, Soy Lecithin 5g/L, Polysorbate 20 40mL/L
For Use With (Application) Sterility and non-sterile product testing
Packaging Type Bottle
Quantity 90 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.