Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD CBA Human Chemokine Kit

Catalog No. bdb552990 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
80 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB552990 80 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB552990 Supplier BD Supplier No. 552990
Only null left
Add to Cart
Add to Cart

Includes

Reagents for capture and detection of IL-8, RANTES, MIG, MCP-1, and IP-10, wash buffer, assay diluent, cytometer setup reagents

Optimized for analysis of specific chemokines in tissue culture supernatants, EDTA-treated plasma (EDTA-plasma), and serum samples

  • Uses the sensitivity of amplified fluorescence detection by flow cytometry to measure soluble analytes in a particle-based immunoassay
  • Each bead in the kit provides a capture surface for a specific protein and is analogous to an individually coated well in an ELISA plate
  • The Capture Bead mixture is in suspension to allow for the detection of multiple analytes in a small volume sample
  • The broad dynamic range of fluorescent detection via flow cytometry and the efficient capturing of analytes via suspended particles enable assay to use fewer sample dilutions and to determine the concentration of an unknown analyte in substantially less time (compared to conventional ELISA)
  • Can be used to quantitatively measure Interleukin-8 (CXCL8/IL-8), RANTES (CCL5/RANTES), Monokine-induced by Interferon-γ (CXCL9/MIG), Monocyte Chemoattractant Protein-1 (CCL2/MCP-1), and Interferon-γ-induced Protein-10 (CXCL10/IP-10) levels in a single sample
  • Provides sufficient reagents for the quantitative analysis of 80 samples

For research use only. Not for use in diagnostic or therapeutic procedures.

TRUSTED_SUSTAINABILITY

Specifications

Reactivity Human
Detects IL-8, RANTES, MIG, MCP-1 and IP-10
For Use With (Application) For analysis of specific chemokines in Tissue Culture supernatants, EDTA-treated Plasma (EDTA-plasma) and Serum samples
Quantity 80 Tests
Content And Storage 2°C to 8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.