Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD CBA Human Th1/Th2 Cytokine Kit

Catalog No. bdb550749 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
80 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB550749 80 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB550749 Supplier BD Supplier No. 550749
Only null left
Add to Cart
Add to Cart

Includes

Reagents for capture and detection of IL-2, IL-4, IL-5, IL-10, TNF, and IFN-γ, wash buffer, assay diluent, serum enhancement buffer, cytometer setup reagents

Optimized for analysis of specific cytokines in tissue culture supernatants, EDTA plasma, and serum samples

BD CBA assays provide a method of capturing a soluble analyte or set of analytes with beads of known size and fluorescence, making it possible to detect analytes using flow cytometry.

  • Can be used to quantitatively measure Interleukin-2 (IL-2), Interleukin-4 (IL-4), Interleukin-5 (IL-5), Interleukin-10 (IL-10), Tumor Necrosis Factor (TNF), and Interferon-γ (IFN-γ) protein levels in a single sample
  • Detection reagent provided is a mixture of phycoerythrin (PE)-conjugated antibodies, which provides a fluorescent signal in proportion to the amount of bound analyte
  • Provides sufficient reagents for analysis of 80 samples

For research use only. Not for use in diagnostic or therapeutic procedures.

TRUSTED_SUSTAINABILITY

Specifications

Reactivity Human
Detects IL-2, IL-4, IL-5, IL-10, TNF and IFN-g
For Use With (Application) For analysis of specific cytokines in Tissue Culture supernatants, EDTA Plasma and Serum samples
Quantity 80 Tests
Content And Storage 2°C to 8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.