Learn More
Description
Includes
Reagents for capture and detection of IL-2, IL-4, IL-5, IL-6, TNF, and IFN-γ; wash buffer; assay diluent; cytometer setup reagents
BD CBA assays provide a method of capturing a soluble analyte or set of analytes with beads of known size and fluorescence, making it possible to detect analytes using flow cytometry.
- Can be used to quantitatively measure Interleukin-2 (IL-2), Interleukin-4 (IL-4), Interleukin-5 (IL-5), Interleukin-6 (IL-6), Tumor Necrosis Factor (TNF), and Interferon-γ (IFN-γ) protein levels in a single sample
- Antibodies are human-specific and the standards are recombinant human proteins
- Antibodies are cross-reactive with non-human primate cytokines
- Kit uses recombinant human proteins and human-specific antibodies for the measurement of NHP proteins, but measured relative values are reported in pg/mL
- Provides sufficient reagents for 80 tests
For research use only. Not for use in diagnostic or therapeutic procedures.
Specifications
Specifications
| Reactivity | Baboon, Cynomolgus, Rhesus |
| Detects | IL-2, IL-4, IL-5, IL-6, TNF and IFN-g |
| For Use With (Application) | For analysis of specific Cytokines in Tissue Culture supernatants, EDTA Plasma and Serum Samples from rhesus, cynomolgus, baboon or pigtailed macaque non-Human primate species |
| Quantity | 80 Tests |
| Content And Storage | 2°C to 8°C |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.